Cyluc1 für die In-vivo-Bildgebung
Lieferant: AAT BIOQUEST
Achtung
Synonyme: (4S)-2-(6,7-Dihydro-5H-thiazolo[4,5-f]indol-2-yl)-4,5-dihydro-thiazole-4-carboxylic acid, (S)-2-(6,7-Dihydro-5H-thiazolo[4,5-f]indol-2-yl)-4,5-dihydrothiazole-4-carboxylic acid, CycLuc1
12480 12481 12482
AATB12480EA 152 EUR
AATB12480 AATB12481 AATB12482
Cyluc1 für die In-vivo-Bildgebung
Cyluc1
Cyluc1 is a synthetic cyclic alkylaminoluciferin that exhibits properties that are superior to those of D-luciferin for in vivo imaging due to its red-shifted luminescence.
  • High-affinity luciferase binding
  • Significantly higher light output
  • 10X brighter signal than D-luciferin
Molecular Weight: 305,37 g/mol
MDL: MFCD28133582
CAS-Nummer: 1247879-16-8
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0ce9ab8abd4e49b9134731913f6403775e4d826d&vid=698626d3b453490cc3400ba4e79e8f3b0a2ff667&prid=web&clid=SAPDAM