BC-NH2 [2-((4-(Aminomethyl)benzyl)oxy)pyrimidin-4-amine]

Lieferant: AAT BIOQUEST
Achtung

12562 12563
AATB12562EA 292 EUR
AATB12562 AATB12563
BC-NH2 [2-((4-(Aminomethyl)benzyl)oxy)pyrimidin-4-amine]
BC-NH2 [2-((4-(Aminomethyl)benzyl)oxy)pyrimidin-4-amine]

CLIP-tag is a modified version of SNAP-tag.


  • CLIP tag building block
  • High quality
  • Multiple applications

Formel: C₁₂H₁₄N₄O
MDL: MFCD31568524
CAS-Nummer: 1004524-64-4
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=039cdbc57b838b2bc6c6b00e2daafac66598f907&vid=4962292d57dc73e1d2fb09c12019a989032686e8&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR