DAF-2 diacetate zur Stickoxiderkennung
Lieferant: AAT BIOQUEST
Achtung
Synonyme: 2-​(3,​6-​diacetyloxy-​4,​5-​diamino-​9H-​xanthen-​9-​yl)-​benzoic acid, DAF-2 DA, 4,5-Diaminofluorescein diacetate
16293 16294
AATB16293EA 292 EUR
AATB16293 AATB16294
DAF-2 diacetate zur Stickoxiderkennung
DAF-2 diacetate
DAF-2 diacetate is a more cell-permeable version of DAF-2, a sensitive fluorescent indicator for the detection and bioimaging of nitric oxide (NO).
  • Sensitive NO detection
Formel: C₂₄H₁₈N₂O₇
Molecular Weight: 446,41 g/mol
MDL: MFCD05865101
CAS-Nummer: 205391-02-2
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dfd9e28eb531c9992e58c1bdf8ed92c2c5f221e1&vid=d7ccd311dd7e94ba6178a7a48a58cf45d84b932b&prid=web&clid=SAPDAM