Aminoallyl-dUTP sodium salt (aa-dUTP) 4 mM in TRIS Puffer ≥95% pH 7.5
Lieferant: AAT BIOQUEST
Achtung
Synonyme: AA-dUTP, 5-[3-Aminoallyl]-2′-deoxyuridine 5′-triphosphate
17004 17005
AATB17004EA 152 EUR
AATB17004 AATB17005
Aminoallyl-dUTP sodium salt (aa-dUTP) 4 mM in TRIS Puffer ≥95% pH 7.5
Aminoallyl-dUTP sodium salt (aa-dUTP)
The amine-modified deoxyuridine 5'-triphosphates (such as aminoallyl-dUTP) can be used to produce amine-containing DNA by conventional enzymatic incorporation methods such as reverse transcription, nick translation, random primed labeling, or PCR.
Formel: C₁₂H₂₀N₃O₁₄P₃·xNa+
Molecular Weight: 523.22 g/mol (free acid)
CAS-Nummer: 936327-10-5
ADR: 5 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=725c9fffa112cc3bc13da7b45361898fc514fbef&vid=a47e51d3224b1e3e4bf2c38b7a3650aa99eca628&prid=web&clid=SAPDAM