Cal-590™ AM

Lieferant: AAT BIOQUEST
Achtung

20510 20511 20512
AATB20510EA 307 EUR
AATB20510 AATB20511 AATB20512
Cal-590™ AM
Cal-590™ AM

Calcium measurement is critical for numerous biological investigations.


  • Improved sensitivity
  • Improved calcium response
  • Long-wavelength emission


Bitte beachten For research use only.

Molecular Weight: 1266.81 g/mol
ADR: 5 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR