Cal Red™ R525/650 AM
Lieferant: AAT BIOQUEST
Achtung
20590 20591
AATB20590EA 769 EUR
AATB20590 AATB20591
Cal Red™ R525/650 AM
Cal Red™ R525/650 AM
The intracellular calcium flux assay is a widely used method for monitoring the activities of GPCRs and calcium channels.
  • Calcium-dependent fluorescence
  • Improved sensitivity
  • Compatible with hts filters
  • Ratiometric detection
  • Cell permeant
Bitte beachten For research use only.
ADR: 5,II