Calbryte™ 590 AM

Lieferant: AAT BIOQUEST
Achtung

20700 20701 20702
AATB20700EA 307 EUR
AATB20700 AATB20701 AATB20702
Calbryte™ 590 AM
Calbryte™ 590 AM

The intracellular calcium flux assay is a widely used method in monitoring signal transduction pathways and high throughput screening of G protein coupled receptors (GPCRs) and calcium channel targets.


  • Improved background/signal ratio
  • Excellent intracellular retention
  • Long-wavelength emission
  • Cell permeant


Bitte beachten For research use only.

Molecular Weight: 1218.77 g/mol
ADR: 5 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR