NAADP-AM
Lieferant: AAT BIOQUEST
|
Achtung
|
20997
20998
21000
AATB20997EA
460
EUR
AATB20997
AATB20998
AATB21000
NAADP-AM
NAADP-AM
NAADP (nicotinic acid adenine dinucleotide phosphate) is a recently discovered second messenger.
- Well retained in cells
- Cell permeable
- Multiple applications
Bitte beachten For research use only.
Molecular Weight:
1032.64 g/mol |
ADR:
5 II |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...