Fura-FF, pentapotassium salt [Fura-2FF, pentapotassium salt] ≥95%
Lieferant: AAT BIOQUEST
|
Achtung
|
21028
21029
AATB21028EA
307
EUR
AATB21028
AATB21029
Fura-FF, pentapotassium salt [Fura-2FF, pentapotassium salt] ≥95%
Fura-FF, pentapotassium salt [Fura-2FF, pentapotassium salt]
Among the ratiometric calcium indicators, Fura-2 and Indo-1 are most commonly used.
Formel:
C₂₈H₁₈F₂K₅N₃O₁₄ Molecular Weight: 853,94 g/mol |
CAS-Nummer:
192140-58-2 ADR: 5,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...