Rhod-4™ AM
Lieferant: AAT BIOQUEST
|
Achtung
|
21120
21121
21122
21123
AATB21120EA
769
EUR
AATB21120
AATB21121
AATB21122
AATB21123
Rhod-4™ AM
Rhod-4™ AM
Calcium measurement is critical for numerous biological investigations.
- Well-cited
- Long-wavelength emission
- Cell permeant
Bitte beachten For research use only.
Molecular Weight:
1015.96 g/mol |
ADR:
5 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...