Cal-520® AM
Lieferant: AAT BIOQUEST
|
Achtung
|
21130
21131
AATB21130EA
307
EUR
AATB21130
AATB21131
Cal-520® AM
Cal-520® AM
Cal-520® AM provides a robust homogeneous fluorescence-based assay tool for detecting intracellular calcium mobilization.
- Improved background/signal ratio
- Suitable for all major fluorescence platforms
- Cell permeant
Bitte beachten For research use only.
Molecular Weight:
1102.95 g/mol |
ADR:
5 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...