Cal-520® potassium salt
Lieferant: AAT BIOQUEST
|
Achtung
|
21140
21141
AATB21140EA
460
EUR
AATB21140
AATB21141
Cal-520® potassium salt
Cal-520® potassium salt
Cal-520® provides a robust homogeneous fluorescence-based assay tool for detecting intracellular calcium mobilization.
- Improved background/signal ratio
- Suitable for all major fluorescence platforms
- Cell impermeant
Bitte beachten For research use only.
Molecular Weight:
921,08 g/mol |
ADR:
5,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...