Coelenterazine hcp ≥95%
Lieferant: AAT BIOQUEST
|
Achtung
|
21154
21160
AATB21154EA
152
EUR
AATB21154
AATB21160
Coelenterazine hcp ≥95%
Coelenterazine hcp
Calcium measurement is critical for numerous biological investigations.
Formel:
C₂₅H₂₅N₃O₂ Molecular Weight: 399,48 g/mol |
MDL:
MFCD03453204 CAS-Nummer: 123437-32-1 ADR: 5,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...