5(6)-Carboxytetramethylrhodamine succinimidyl ester [5(6)-TAMRA SE] ≥95%

Lieferant: AAT BIOQUEST
Achtung

Synonyme: (5(6)-TAMRA SE

370 371 372
AATB370EA 152 EUR
AATB370 AATB371 AATB372
5(6)-Carboxytetramethylrhodamine succinimidyl ester [5(6)-TAMRA SE] ≥95%
5(6)-Carboxytetramethylrhodamine succinimidyl ester [5(6)-TAMRA SE]

The succinimidyl esters of 5-TAMRA, 6-TAMRA or the mixed isomers are the primary labeling reagents for the preparation of orange fluorescent bioconjugates, including peptide, protein, nucleotide and nucleic acid conjugates, especially fluorescent antibodies and avidin derivatives used in immunochemistry.

Formel: C₂₉H₂₅N₃O₇
Molecular Weight: 527.52 g/mol
MDL: MFCD00077324
CAS-Nummer: 150408-83-6
ADR: 5 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2170a7b242e001cec4beadef49801e7c36812f37&vid=e59b08f03e44ff31fbab8cb46d13c7c8a2d20e40&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR