6-Carboxytetramethylrhodamine succinimidyl ester (6-TAMRA SE) ≥90%
Lieferant: AAT BIOQUEST
|
Achtung
|
Synonyme:
6-Carboxytetramethylrhodamine N-succinimidyl ester, 6-TAMRA, SE, 6-TAMRA N-succinimidyl ester
376
377
378
AATB376EA
159
EUR
AATB376
AATB377
AATB378
6-Carboxytetramethylrhodamine succinimidyl ester (6-TAMRA SE) ≥90%
6-Carboxytetramethylrhodamine succinimidyl ester (6-TAMRA SE)
The single TAMRA isomers are increasingly preferred for labeling peptides and nucleotides because they give better resolution in HPLC purification that is often required in the conjugation processes.
Formel:
C₂₉H₂₅N₃O₇ Molecular Weight: 527.52 g/mol |
CAS-Nummer:
150810-69-8 ADR: 5 II |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...