Tetramethylrhodamine 6-isothiocyanate chloride ≥90%
Lieferant: AAT BIOQUEST
Achtung
Synonyme: 6-TRITC
417 418
AATB417EA 307 EUR
AATB417 AATB418
Tetramethylrhodamine 6-isothiocyanate chloride ≥90%
Tetramethylrhodamine 6-isothiocyanate chloride (6-TRITC)
6-TRITC (also called R isomer) is the other isomer of the TRITC labeling reagent that is widely used in preparing bioconjugates of proteins and nucleic acids.
Formel: C₂₅H₂₂ClN₃O₃S
Molecular Weight: 479,98 g/mol
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4c25a9ea56b575f59350a2a368f8779c4e033485&vid=b9dbf62776c36a6d4faf443deb24fdc871275cb6&prid=web&clid=SAPDAM