FMOC-Glu(biotinyl-PEG)-OH
Lieferant: AAT BIOQUEST
Achtung
Synonyme: Fmoc-L-Glu(biotinyl-PEG)-OH, (S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-5,21-dioxo-25-((3aS,4S,6aR)-2-oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)-10,13,16-trioxa-6,20-diazapentacosanoic acid
5026 5027
AATB5026EA 307 EUR
AATB5026 AATB5027
FMOC-Glu(biotinyl-PEG)-OH
FMOC-Glu(biotinyl-PEG)-OH
This PEG biotin-labeled amino acid has excellent solubility in DMF and other solvents used in SPPS.
  • Specific reactivity
  • Stable conjugation
  • High solubility
Molecular Weight: 797,96 g/mol
MDL: MFCD18782826
CAS-Nummer: 817169-73-6
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bcc02031909f54c4ada87e3a3c7750ff4d934ce8&vid=5411f68c456ed0bfb9f2a2644fab433b93621a0a&prid=web&clid=SAPDAM