Phalloidin lysine
Lieferant: AAT BIOQUEST
|
Achtung
|
5304
5305
AATB5304EA
137
EUR
AATB5304
AATB5305
Phalloidin lysine
Phalloidin lysine
Phalloidin lysine is a convenient building block that can be used for developing a variety of phalloidin derivatives that may be for studying cellular structures and other biological applications.
- Useful in live or fixed cells
- High affinity for F-actin
ADR:
5,II |