Saponin
Lieferant: Thermo Fisher Scientific
|
Achtung
|
A18820.14
A18820.22
A18820.14EA
47.4
EUR
A18820.14
A18820.22
Saponin
Saponin
CAS No.: 8047-15-2
Physical Form: Powder
Appearance: White to pale yellow
Melting Point: 158° dec.
Merck Reference: 14,8365
MDL No.: MFCD00081981
A glycoside used to permeabilize cellular membranes
Schmelzpunkt:
158 °C (Zers.) Dichte: 1.015…1.02 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00081981 CAS-Nummer: 8047-15-2 EINECS: 232-462-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...