Saponin

Lieferant: Serva

34655.01 34655.02
SERA34655.01EA 56.6 EUR
SERA34655.01 SERA34655.02
Saponin
Saponin
Dichte: 1.015…1.02 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00081981
CAS-Nummer: 8047-15-2
EINECS: 232-462-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR