Gibberellinsäure ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
GA3, Gibberellin A3, (3S,3aS,4S,4aS,6S,8aR,8bR,11S)-6,11-Dihydroxy-3-methyl-12-methylen-2-oxo-4a,6-ethano-3,8b-prop-1-enoperhydroindeno[1,2-b]furan-4-carbonsäure
45569.04
45569.09
45569.ME
45569.04EA
82
EUR
45569.04
45569.09
45569.ME
Gibberellinsäure ≥98%
Gibberellic acid
Purity: 98%
CAS No.: 77-06-5
Molecular Formula: C19H22O6
Formula Weight: 346.37
Physical Form: Powder
Appearance: White to off-white
Melting Point: 222° dec.
Beilstein Reference: 54346
Merck Reference: 14,4419
Optical Rotation: +78° (c=2 in methanol)
MDL No.: MFCD00079329
Plant growth hormone
Formel:
C₁₉H₂₂O₆ Molecular Weight: 346.38 g/mol Schmelzpunkt: 222 °C (Zers.) Dichte: 1.386 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00079329 CAS-Nummer: 77-06-5 EINECS: 201-001-0 Merck Index: 14,4419 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...