(±)-4-Brommandelsäure ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
(±)-2-(4-Bromphenyl)-2-hydroxyessigsäure, (±)-4-Bromphenylglykolsäure, (±)-2-(4-Bromphenyl)-2-hydroxyethansäure
A12978.06
A12978.14
A12978.22
A12978.06EA
40.6
EUR
A12978.06
A12978.14
A12978.22
(±)-4-Brommandelsäure ≥98%
(±)-4-Brommandelsäure
Purity: 98+%
CAS No.: 6940-50-7
Molecular Formula: C8H7BrO3
Formula Weight: 231.05
Melting Point: 115-119°
Beilstein Reference: 1567909
Merck Reference: 14,1424
MDL No.: MFCD00004232
Formel:
C₈H₇BrO₃ Molecular Weight: 231,05 g/mol Siedepunkt: 344 °C (1013 hPa) Schmelzpunkt: 115…119 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004232 CAS-Nummer: 6940-50-7 EINECS: 230-085-1 Merck Index: 14,1424 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...