(±)-4-Brommandelsäure

Lieferant: FLUOROCHEM

Synonyme: (±)-2-(4-Bromphenyl)-2-hydroxyessigsäure, (±)-4-Bromphenylglykolsäure, (±)-2-(4-Bromphenyl)-2-hydroxyethansäure

019427-1G 019427-5G
FLUO019427-1GEA 0 EUR
FLUO019427-1G FLUO019427-5G
(±)-4-Brommandelsäure
(±)-4-Brommandelsäure
Formel: C₈H₇BrO₃
Molecular Weight: 231,05 g/mol
Siedepunkt: 344 °C (1013 hPa)
Schmelzpunkt: 115…118 °C
MDL: MFCD00004232
CAS-Nummer: 6940-50-7
EINECS: 230-085-1
Merck Index: 13,01409

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR