(±)-2-Chlormandelsäure ≥98%
Lieferant: Thermo Scientific
Synonyme:
(±)-2-Chlorphenylglykolsäure, (±)-2-(2-Chlorphenyl)-2-hydroxyessigsäure, (±)-2-(2-Chlorphenyl)-2-hydroxyethansäure
B21525.06
B21525.14
B21525.06EA
34.6
EUR
B21525.06
B21525.14
(±)-2-Chlormandelsäure ≥98%
(±)-2-Chlormandelsäure
Purity: 98%
CAS No.: 10421-85-9
Molecular Formula: C8H7ClO3
Formula Weight: 186.60
Melting Point: 80-85°
Beilstein Reference: 2367070
Formel:
C₈H₇ClO₃ Molecular Weight: 186,59 g/mol Siedepunkt: 350 °C (1013 hPa) Schmelzpunkt: 80…85 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00084962 CAS-Nummer: 10421-85-9 EINECS: 233-900-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...