(±)-2,4-Difluormandelsäure

Lieferant: FLUOROCHEM

Synonyme: (±)-2,4-Difluorophenylglycolic acid

005209-5G 005209-1G 005209-25G
FLUO005209-5GEA 281 EUR
FLUO005209-5G FLUO005209-1G FLUO005209-25G
(±)-2,4-Difluormandelsäure
(±)-2,4-Difluormandelsäure
Formel: C₈H₆F₂O₃
Molecular Weight: 188,13 g/mol
MDL: MFCD00061296
CAS-Nummer: 132741-30-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR