(±)-2,6-Difluormandelsäure

Lieferant: FLUOROCHEM

Synonyme: (±)-2-(2,6-Difluorphenyl)-2-hydroxyessigsäure, (±)-2,6-Difluorphenylglykolsäure, (±)-2-(2,6-Difluorphenyl)-2-hydroxyethansäure

004824-1G 004824-5G
FLUO004824-1GEA 57.9 EUR
FLUO004824-1G FLUO004824-5G
(±)-2,6-Difluormandelsäure
(±)-2,6-Difluormandelsäure
Formel: C₈H₆F₂O₃
Molecular Weight: 188,13 g/mol
MDL: MFCD00061298
CAS-Nummer: 207981-50-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR