(±)-3,4-Difluormandelsäure

Lieferant: FLUOROCHEM

Synonyme: (±)-3,4-Difluorophenylglycolic acid

005212-1G 005212-5G 005212-25G
FLUO005212-1GEA 48.8 EUR
FLUO005212-1G FLUO005212-5G FLUO005212-25G
(±)-3,4-Difluormandelsäure
(±)-3,4-Difluormandelsäure
Formel: C₈H₆F₂O₃
Molecular Weight: 188,13 g/mol
MDL: MFCD00061299
CAS-Nummer: 132741-29-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR