(±)-4-(Trifluormethyl)mandelsäure

Lieferant: FLUOROCHEM

Synonyme: (±)-4-(Trifluormethyl)phenylglykolsäure, (±)-2-(4-(Trifluormethyl)phenyl)-2-hydroxyethansäure, (±)-2-(4-(Trifluormethyl)phenyl)-2-hydroxyessigsäure

007628-1G 007628-5G 007628-25G
FLUO007628-1GEA 0 EUR
FLUO007628-1G FLUO007628-5G FLUO007628-25G
(±)-4-(Trifluormethyl)mandelsäure
(±)-4-(Trifluormethyl)mandelsäure
Formel: C₉H₇F₃O₃
Molecular Weight: 220.15 g/mol
Siedepunkt: 314
Schmelzpunkt: 129…130.5 °C
MDL: MFCD00009724
CAS-Nummer: 395-35-7
EINECS: 206-900-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR