(±)-4-Fluormandelsäure ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
(±)-2-(4-Fluorphenyl)-2-hydroxyessigsäure, (±)-2-(4-Fluorphenyl)-2-hydroxyethansäure, (±)-4-Fluorphenylglykolsäure
A13391.03
A13391.06
A13391.14
A13391.03EA
16.4
EUR
A13391.03
A13391.06
A13391.14
(±)-4-Fluormandelsäure ≥98%
(±)-4-Fluormandelsäure
Purity: 98%
CAS No.: 395-33-5
Molecular Formula: C8H7FO3
Formula Weight: 170.14
Melting Point: 133-138°
Beilstein Reference: 2211001
Formel:
C₈H₇FO₃ Molecular Weight: 170,14 g/mol Schmelzpunkt: 133…138 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00044402 CAS-Nummer: 395-33-5 EINECS: 206-899-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...