DL-Prolin, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
(±)-Prolin, (±)-1,3,4,5-Tetrahydro-2H-pyrrol- 2-carbonsäure, (±)-Pyrrolidin-2-carbonsäure, (±)-Azolidin-2-carbonsäure, H-DL-Pro-OH
M-Clarity™ Qualitätsstufe = MQ100
171824-1G
171824-5G
SIAL171824-1GEA
31
EUR
SIAL171824-1G
SIAL171824-5G
DL-Prolin, Sigma-Aldrich®
DL-Prolin
Formel:
C₅H₉NO₂ Molecular Weight: 115.13 g/mol Siedepunkt: 245 Schmelzpunkt: 208…210 °C Dichte: 1.38 g/cm³ (22.9 °C) |
MDL:
MFCD00005250 CAS-Nummer: 609-36-9 EINECS: 210-189-3 Merck Index: 12,07963 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...