4-Brom-α-methylbenzylalkohol
Lieferant: FLUOROCHEM
Synonyme:
4-Bromophenyl methyl carbinol, 1-(4-Bromophenyl)ethanol, p-Bromo-α-methylbenzyl alcohol
001129-100G
001129-25G
FLUO001129-100GEA
254
EUR
FLUO001129-100G
FLUO001129-25G
4-Brom-α-methylbenzylalkohol
4-Brom-α-methylbenzylalkohol
Formel:
C₈H₉BrO Siedepunkt: 253 °C (1013 hPa) Dichte: 1,469 g/cm³ (15.2 °C) |
MDL:
MFCD00004514 CAS-Nummer: 5391-88-8 EINECS: 226-389-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...