4-Chlor-α-methylbenzylalkohol
Lieferant: FLUOROCHEM
Synonyme:
1-(4-Chlorophenyl)ethanol, 4-Chlorophenyl methyl carbinol, p-Chloro-α-methylbenzyl alcohol
002971-5G
002971-25G
FLUO002971-5GEA
48.8
EUR
FLUO002971-5G
FLUO002971-25G
4-Chlor-α-methylbenzylalkohol
4-Chlor-α-methylbenzylalkohol
Formel:
C₈H₉ClO Molecular Weight: 156,61 g/mol Dichte: 1,17 g/cm³ (20 °C) |
MDL:
MFCD00041038 CAS-Nummer: 3391-10-4 EINECS: 222-223-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...