2-Fluor-α-methylbenzylalkohol

Lieferant: FLUOROCHEM

006337-1G 006337-5G 006337-10G 006337-50G
FLUO006337-1GEA 0 EUR
FLUO006337-1G FLUO006337-5G FLUO006337-10G FLUO006337-50G
2-Fluor-α-methylbenzylalkohol
2-Fluor-α-methylbenzylalkohol
Formel: C₈H₉FO
Molecular Weight: 140.16 g/mol
MDL: MFCD00014401
CAS-Nummer: 445-26-1
EINECS: 207-155-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR