3-Amino-α-methylbenzylalkohol 98%
Lieferant: Thermo Scientific
Achtung
|
Synonyme:
m-Amino-α-methylbenzylalkohol, 3-Aminophenyl-methylcarbinol, 1-(3-Aminophenyl)ethanol, 3-(1-Hydroxyethyl)anilin
L06513.14
L06513.22
L06513.14EA
45.9
EUR
L06513.14
L06513.22
3-Amino-α-methylbenzylalkohol 98%
3-Amino-α-methylbenzylalkohol
Purity: 98%
CAS No.: 2454-37-7
Molecular Formula: C8H11NO
Formula Weight: 137.18
Flash Point: 157°(314°F)
Melting Point: 66-69°
Boiling Point: 217°
Beilstein Reference: 2802415
MDL No.: MFCD00007798
Formel:
C₈H₁₁NO Molecular Weight: 137,18 g/mol Siedepunkt: 217 °C Schmelzpunkt: 66…69 °C Flammpunkt: 157 °C (314 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007798 CAS-Nummer: 2454-37-7 EINECS: 219-525-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...