(±)-2-Phenylpiperazin 96%

Lieferant: Thermo Scientific
Gefahr

H26502.06 H26502.14
H26502.06EA 108 EUR
H26502.06 H26502.14
(±)-2-Phenylpiperazin 96%
(±)-2-Phenylpiperazin

Purity: 96% CAS No.: 5271-26-1 Molecular Formula: C10H14N2 Formula Weight: 162.24 Melting Point: 83-87° UN No.: 3259 MDL No.: MFCD01871362

Formel: C₁₀H₁₄N₂
Molecular Weight: 162,23 g/mol
Schmelzpunkt: 83…87 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01871362
CAS-Nummer: 5271-26-1
EINECS: None
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR