(±)-2-Phenylpiperazin
Lieferant: FLUOROCHEM
021555-5G
021555-1G
FLUO021555-5GEA
294
EUR
FLUO021555-5G
FLUO021555-1G
(±)-2-Phenylpiperazin
(±)-2-Phenylpiperazin
Formel:
C₁₀H₁₄N₂ Molecular Weight: 162,23 g/mol |
MDL:
MFCD01871362 CAS-Nummer: 5271-26-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...