3-Chlor-4-nitroanilin 97%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 4-Amino-2-chloronitrobenzene

OR2640-5G OR2640-25G OR2640-100G
APOSOR2640-5GEA 21.9 EUR
APOSOR2640-5G APOSOR2640-25G APOSOR2640-100G
3-Chlor-4-nitroanilin 97%
3-Chloro-4-nitroaniline

Delivery of this item to mainland UK is £40, additional charge will be added at order confirmation stage by our sales team.

Formel: C6H5ClN2O2
Schmelzpunkt: 157 - 161 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00085922
CAS-Nummer: 825-41-2
UN: 2811
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR