4-Ethoxy-4'-methylbenzophenone
Lieferant: Apollo Scientific
|
Achtung
|
OR52285-5G
OR52285-25G
OR52285-100G
APOSOR52285-5GEA
26.1
EUR
APOSOR52285-5G
APOSOR52285-25G
APOSOR52285-100G
4-Ethoxy-4'-methylbenzophenone
4-Ethoxy-4'-methylbenzophenone
Formel:
C₁₆H₁₆O₂ |
MDL:
MFCD03138264 CAS-Nummer: 65629-84-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...