2-Methoxypropen

Lieferant: Apollo Scientific
Gefahr

Synonyme: 2-Methoxy-1-propene, Isopropenyl methyl ether, 2-Methoxyprop-1-ene

OR17619-5G OR17619-25G OR17619-100G
APOSOR17619-5GEA 50.9 EUR
APOSOR17619-5G APOSOR17619-25G APOSOR17619-100G
2-Methoxypropen
2-Methoxypropene

Delivery of this item to mainland UK is £40, additional charge will be added at order confirmation stage by our sales team.

Formel: C₄H₈O
Molecular Weight: 72,11 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00014929
CAS-Nummer: 116-11-0
UN: 1993
ADR: 3,I

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR