4-Chloro-2-(trifluoromethyl)nicotinic acid 95%
Lieferant: Apollo Scientific
Achtung
Synonyme: 4-Chloro-2-(trifluoromethyl)pyridine-3-carboxylic acid, 3-Carboxy-4-chloro-2-(trifluoromethyl)pyridine
PC49634-250MG PC49634-1G PC49634-5G
APOSPC49634-250MGEA 144 EUR
APOSPC49634-250MG APOSPC49634-1G APOSPC49634-5G
4-Chloro-2-(trifluoromethyl)nicotinic acid 95%
4-Chloro-2-(trifluoromethyl)nicotinic acid
Formel: C₇H₃ClF₃NO₂
Molecular Weight: 225.55 g/mol
MDL: MFCD22546902
CAS-Nummer: 1018678-39-1