Ethyl-5-chlor-1H-indol-2-carboxylat

Lieferant: Apollo Scientific
Achtung

Synonyme: Ethyl 5-chloro-1H-indole-2-carboxylate, Ethyl 5-chloro-2-indolecarboxylate

OR10934-25G OR10934-100G
APOSOR10934-25GEA 74.1 EUR
APOSOR10934-25G APOSOR10934-100G
Ethyl-5-chlor-1H-indol-2-carboxylat
Ethyl-5-chloro-1H-indole-2-carboxylate
Formel: C₁₁H₁₀ClNO₂
Molecular Weight: 223.66 g/mol
Schmelzpunkt: 166…168 °C
MDL: MFCD00005610
CAS-Nummer: 4792-67-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR