5-Brom-2-(trifluormethyl)benzoesäure

Lieferant: Apollo Scientific
Achtung

PC250010-1G PC250010-5G
APOSPC250010-1GEA 130 EUR
APOSPC250010-1G APOSPC250010-5G
5-Brom-2-(trifluormethyl)benzoesäure
5-Brom-2-(trifluormethyl)benzoesäure
Formel: C₈H₄BrF₃O₂
Schmelzpunkt: 88 - 89 °C
MDL: MFCD12026306
CAS-Nummer: 654-97-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR