Methyl-4-fluor-3-methylbenzoat

Lieferant: Apollo Scientific
Achtung

Synonyme: Methyl 4-fluoro-3-methylbenzoate

PC3446-10G PC3446-25G
APOSPC3446-10GEA 82.8 EUR
APOSPC3446-10G APOSPC3446-25G
Methyl-4-fluor-3-methylbenzoat
Methyl-4-fluor-3-methylbenzoat
Formel: C₉H₉FO₂
Molecular Weight: 168.1650 g/mol
MDL: MFCD06203783
CAS-Nummer: 180636-50-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR