3-Brom-5-chlor-2-iodbenzonitril 95%
Lieferant: Apollo Scientific
|
Gefahr
|
OR61187-1G
OR61187-5G
OR61187-10G
APOSOR61187-1GEA
250
EUR
APOSOR61187-1G
APOSOR61187-5G
APOSOR61187-10G
3-Brom-5-chlor-2-iodbenzonitril 95%
3-Brom-5-chlor-2-iodbenzonitril
Formel:
C₇H₂BrClIN Molecular Weight: 342.3600 g/mol |
MDL:
MFCD11846071 CAS-Nummer: 1160574-37-7 UN: 3439 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...