2-N-methylamino-6-bromobenzoic acid 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Bromo-6-(methylamino)benzoic acid

OR71110-250MG OR71110-1G
APOSOR71110-250MGEA 125 EUR
APOSOR71110-250MG APOSOR71110-1G
2-N-methylamino-6-bromobenzoic acid 95%
2-N-methylamino-6-bromobenzoic acid
Formel: C₈H₈BrNO₂
Molecular Weight: 230,06 g/mol
MDL: MFCD28356796
CAS-Nummer: 1369866-00-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR