1-(5-Bromo-3-chloro-2-fluorophenyl)ethanone 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5'-Bromo-3'-chloro-2'-fluoroacetophenone, 1-(5-Bromo-3-chloro-2-fluorophenyl)ethan-1-one
PC37883-250MG
PC37883-1G
PC37883-5G
APOSPC37883-250MGEA
190
EUR
APOSPC37883-250MG
APOSPC37883-1G
APOSPC37883-5G
1-(5-Bromo-3-chloro-2-fluorophenyl)ethanone 95%
1-(5-Bromo-3-chloro-2-fluorophenyl)ethanone
Formel:
C₈H₅BrClFO Molecular Weight: 251.48 g/mol |
MDL:
MFCD26679371 CAS-Nummer: 1536939-67-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...