3-Bromo-4-chloro-2-fluorophenacyl bromide 95%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 2-Bromo-1-(3-Bromo-4-chloro-2-fluorophenyl)ethanone

PC48962-250MG PC48962-1G PC48962-5G
APOSPC48962-250MGEA 219 EUR
APOSPC48962-250MG APOSPC48962-1G APOSPC48962-5G
3-Bromo-4-chloro-2-fluorophenacyl bromide 95%
3-Bromo-4-chloro-2-fluorophenacyl bromide
Formel: C₈H₄Br₂ClFO
MDL: MFCD28733987
CAS-Nummer: 1805478-98-1
UN: 3261
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR