4'-Bromo-2,2,2,2',3'-pentafluoroacetophenone 95%

Lieferant: Apollo Scientific
Gefahr

PC500212-250MG PC500212-1G
APOSPC500212-250MGEA 261 EUR
APOSPC500212-250MG APOSPC500212-1G
4'-Bromo-2,2,2,2',3'-pentafluoroacetophenone 95%
4'-Bromo-2,2,2,2',3'-pentafluoroacetophenone
Formel: C₈H₂BrF₅O
Molecular Weight: 289 g/mol
MDL: MFCD27664852
CAS-Nummer: 1823361-42-7
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR