5-Bromo-2-chloro-4-fluorobenzaldehyde 95%
Lieferant: Apollo Scientific
|
Achtung
|
PC46365-250MG
PC46365-1G
PC46365-5G
APOSPC46365-250MGEA
65.4
EUR
APOSPC46365-250MG
APOSPC46365-1G
APOSPC46365-5G
5-Bromo-2-chloro-4-fluorobenzaldehyde 95%
5-Bromo-2-chloro-4-fluorobenzaldehyde
Formel:
C₇H₃BrClFO Molecular Weight: 237,4535 g/mol |
MDL:
MFCD28652989 CAS-Nummer: 1782815-29-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...