2’-Bromo-3’,6’-difluoroacetophenone 95%
Lieferant: Apollo Scientific
|
Achtung
|
PC501794-250MG
PC501794-1G
PC501794-5G
APOSPC501794-250MGEA
203
EUR
APOSPC501794-250MG
APOSPC501794-1G
APOSPC501794-5G
2’-Bromo-3’,6’-difluoroacetophenone 95%
2’-Bromo-3’,6’-difluoroacetophenone
Formel:
C₈H₅BrF₂O Molecular Weight: 235,03 g/mol |
MDL:
MFCD28061326 CAS-Nummer: 1807029-26-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...